Learn More
Medchemexpress LLC L-Leucine-d7 | 92751-17-2 | 138.22 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN0486S41MG
Encompass_Preferred
L-Leucine-d7 is a deuterium labeled L-Leucine, an essential branched-chain amino acid (BCAA) known to activate the mTOR signaling pathway. This compound serves as a valuable tool in various analytical methods, including quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Activates the mTOR signaling pathway
Catalog No. 50-003-89373