Learn More
Medchemexpress LLC L-leucine, N-(cyclohexylcarbonyl)- | 157116-68-2 | MFCD16377985 | 97.0% | 241.33 g/mol | C13H23NO3 | 250 MG
SDP
Catalog No. 5000209188
Encompass_Preferred
Cyclohexanecarbonyl-L-leucine is an N-cyclohexanecarbonyl derivative of L-leucine used as an amino acid derivative in research. It is supplied as a white to off-white powder (CAS 157116-68-2) with molecular formula C13H23NO3 and molecular weight 241.33 g/mol.
  • Powder form suitable for synthesis and peptide modification.
  • High purity (97.0%) for analytical and research applications.
  • Stable under recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Solvent stability for prepared solutions: -80°C (6 months) or -20°C (1 month).
  • Available in multiple sizes including 250 mg for small-scale experiments.
Catalog No. 50-002-09188 Supplier Medchemexpress LLC Supplier No. 250MGHYW046352
Cyclohexanecarbonyl-L-leucine is an N-cyclohexanecarbonyl derivative of L-leucine used as an amino acid derivative in research. It is supplied as a white to off-white powder (CAS 157116-68-2) with molecular formula C13H23NO3 and molecular weight 241.33 g/mol.
  • Powder form suitable for synthesis and peptide modification.
  • High purity (97.0%) for analytical and research applications.
  • Stable under recommended storage: powder -20°C (3 years) or 4°C (2 years).
  • Solvent stability for prepared solutions: -80°C (6 months) or -20°C (1 month).
  • Available in multiple sizes including 250 mg for small-scale experiments.