Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Leucine, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111014911 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4911 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4911 Supplier MilliporeSigma Supelco Supplier No. PHR11051G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leucine is a non-glucogenic, essential amino acid and a structural component of proteins.

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
TRUSTED_SUSTAINABILITY

Specifications

CAS 61-90-5
Physical Form Neat
Linear Formula (CH3)2 CHCH2CH(NH2)CO2H
MDL Number MFCD00002617
Melting Point >300°C
Synonym (S)-2-Amino-4-methylpentanoic acid
Quantity 1 g
Formula Weight 131.17
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.