Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Leucine tert-butyl ester hydrochloride, 98%

Catalog No. AAH5998006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5998006 5 g
AAH5998003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5998006 Supplier Thermo Scientific Chemicals Supplier No. H5998006
  • $125.40
    $106.65 / Each of 1

    Save $18.75

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2748-02-9 | C3H10NO | 76.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2748-02-9
MDL Number MFCD00038906
ChEBI CHEBI:78502

Specifications

Molecular Formula C3H10NO
Beilstein 3697141
Synonym l-alaninol, s-+-2-amino-1-propanol, s-2-aminopropan-1-ol, 2s-2-aminopropan-1-ol, h-alaninol, alaninol, s-2-amino-1-propanol, +-2-aminopropan-1-ol, 2-aminopropanol, +, unii-v403gh89l1
InChI Key BKMMTJMQCTUHRP-VKHMYHEASA-O
SMILES CC(CO)N
IUPAC Name (2S)-2-aminopropan-1-ol
Quantity 5 g
Molecular Weight (g/mol) 76.12
PubChem CID 80307
Formula Weight 223.74
Optical Rotation +12° (c=2 in DMF)
Percent Purity 98%
Chemical Name or Material L-Leucine tert-butyl ester hydrochloride
Show More Show Less
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.