Learn More
Medchemexpress LLC L-leucinol | 7533-40-6 | MFCD00063676 | 99.5% | 117.19 g/mol | C6H15NO | 50 MG
SDP
Catalog No. 5000300259
Encompass_Preferred
L-Leucinol is the L-enantiomer of leucinol, an amino alcohol used as a competitive aminopeptidase inhibitor and as a building block in peptide synthesis. It is supplied as a high-purity research reagent intended for in vitro enzymatic experiments and synthetic applications.
  • High reported purity (≈99.5%) suitable for research use.
  • Acts as a competitive aminopeptidase inhibitor in enzymatic assays.
  • Usable as a synthetic building block for peptide assembly.
  • Stored under low-temperature conditions for long-term stability.
  • Colorless to light yellow liquid, readily handled in solution.
Catalog No. 50-003-00259 Supplier Medchemexpress LLC Supplier No. HYW01587650MG
L-Leucinol is the L-enantiomer of leucinol, an amino alcohol used as a competitive aminopeptidase inhibitor and as a building block in peptide synthesis. It is supplied as a high-purity research reagent intended for in vitro enzymatic experiments and synthetic applications.
  • High reported purity (≈99.5%) suitable for research use.
  • Acts as a competitive aminopeptidase inhibitor in enzymatic assays.
  • Usable as a synthetic building block for peptide assembly.
  • Stored under low-temperature conditions for long-term stability.
  • Colorless to light yellow liquid, readily handled in solution.