Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-leucinol ((+)-leucinol) | 7533-40-6 | MFCD00063676 | 99.5% | 117.19 g/mol | C6H15NO | 1 ML
SDP

Catalog No. 5000300258
Encompass_Preferred

L-Leucinol ((+)-Leucinol) is an amino alcohol that functions as a competitive aminopeptidase inhibitor. It is used in in vitro enzymatic assays and peptide synthesis, and is supplied both as a solid and as ready-to-use 10 mM solutions in DMSO. Technical specifications include a molecular weight of 117.19 g/mol and high reported purity.

  • Competitive aminopeptidase inhibitor with reported Ki ≈ 17 μM.
  • Suitable for in vitro enzymatic assays and peptide synthesis.
  • Available as 10 mM solution in DMSO and as solid quantities for synthesis.
  • High purity (≈99.5%) for research applications.
  • Molecular weight 117.19 g/mol; formula C6H15NO.
  • Recommended storage: pure form -20°C (long term); in solvent -80°C (long term).

Catalog No. 50-003-00258 Supplier Medchemexpress LLC Supplier No. HYW01587610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Leucinol ((+)-Leucinol) is an amino alcohol that functions as a competitive aminopeptidase inhibitor. It is used in in vitro enzymatic assays and peptide synthesis, and is supplied both as a solid and as ready-to-use 10 mM solutions in DMSO. Technical specifications include a molecular weight of 117.19 g/mol and high reported purity.

  • Competitive aminopeptidase inhibitor with reported Ki ≈ 17 μM.
  • Suitable for in vitro enzymatic assays and peptide synthesis.
  • Available as 10 mM solution in DMSO and as solid quantities for synthesis.
  • High purity (≈99.5%) for research applications.
  • Molecular weight 117.19 g/mol; formula C6H15NO.
  • Recommended storage: pure form -20°C (long term); in solvent -80°C (long term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.