Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Leucyl-4-nitroanilide | 4178-93-2 | 100.0% | 251.28 | 25 G
SDP

Catalog No. 5000451210
Encompass_Preferred

L-Leucyl-4-nitroanilide is a colorimetric substrate used for leucine aminopeptidase assays. This product features:

  • Purity of 99.95%.
  • Appears as a light yellow to yellow solid.
  • Soluble in DMSO at 100 mg/mL.
  • Molecular weight of 251.28.
  • Molecular formula C12H17N3O3.

Catalog No. 50-004-51210 Supplier Medchemexpress LLC Supplier No. 25GHYW011911
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Leucyl-4-nitroanilide is a colorimetric substrate used for leucine aminopeptidase assays. This product features:

  • Purity of 99.95%.
  • Appears as a light yellow to yellow solid.
  • Soluble in DMSO at 100 mg/mL.
  • Molecular weight of 251.28.
  • Molecular formula C12H17N3O3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.