Learn More
Medchemexpress LLC L-Leucyl-4-nitroanilide | 4178-93-2 | >99.95% | 251.29 | 100 MG
SDP
Catalog No. 5000453823
Encompass_Preferred
L-Leucyl-4-nitroanilide is a colorimetric substrate utilized in leucine aminopeptidase assays. This laboratory chemical is intended for research use only, serving as a key substance in various biochemical applications.
  • Stable under recommended storage conditions.
  • Requires handling with appropriate ventilation.
  • Store in a cool, well-ventilated area, away from direct sunlight.
  • Requires personal protective equipment, including eye protection and gloves.
Catalog No. 50-004-53823 Supplier Medchemexpress LLC Supplier No. HYW011911100MG
L-Leucyl-4-nitroanilide is a colorimetric substrate utilized in leucine aminopeptidase assays. This laboratory chemical is intended for research use only, serving as a key substance in various biochemical applications.
  • Stable under recommended storage conditions.
  • Requires handling with appropriate ventilation.
  • Store in a cool, well-ventilated area, away from direct sunlight.
  • Requires personal protective equipment, including eye protection and gloves.