Learn More
Medchemexpress LLC L-Leucyl-4-nitroanilide | 4178-93-2 | MFCD00063681 | 100.0% | 251.28 | 5 G
SDP
Catalog No. 5000367080
Encompass_Preferred
L-Leucyl-4-nitroanilide is a chemical compound primarily used as a colorimetric substrate in leucine aminopeptidase assays.
  • Used as a colorimetric substrate for leucine aminopeptidase assays.
  • Features a molecular formula of C12H17N3O3.
  • Offers an estimated purity of 99.95%.
  • Recommended for storage at room temperature (RT).
  • Certified for research use only.
Catalog No. 50-003-67080 Supplier Medchemexpress LLC Supplier No. HYW0119115G
L-Leucyl-4-nitroanilide is a chemical compound primarily used as a colorimetric substrate in leucine aminopeptidase assays.
  • Used as a colorimetric substrate for leucine aminopeptidase assays.
  • Features a molecular formula of C12H17N3O3.
  • Offers an estimated purity of 99.95%.
  • Recommended for storage at room temperature (RT).
  • Certified for research use only.