Learn More
Medchemexpress LLC L-Leucyl-4-nitroanilide hydrochloride | 16010-98-3 | 100.0% | 287.74 | 250 MG
SDP
Catalog No. 5000367046
Encompass_Preferred
L-Leucyl-4-nitroanilide hydrochloride is a biochemical assay reagent commonly used to measure the activity of enzymes, such as proteases, that cleave peptide bonds. It forms a yellow product, p-nitroaniline, upon cleavage, which is useful for detecting and quantifying protease activity in biological samples like blood or tissue extracts. It is for research use only.
  • Biochemical assay reagent
  • Used as a substrate for enzymes that cleave peptide bonds (e.g., proteases)
  • Cleavage forms a yellow product (p-nitroaniline)
  • Can be used as a biological material or organic compound for life science-related research
  • Appearance: solid
  • Color: off-white to light yellow
  • Useful tool for detecting and quantifying protease activity in biological samples
Catalog No. 50-003-67046 Supplier Medchemexpress LLC Supplier No. 250MGHYW011911
L-Leucyl-4-nitroanilide hydrochloride is a biochemical assay reagent commonly used to measure the activity of enzymes, such as proteases, that cleave peptide bonds. It forms a yellow product, p-nitroaniline, upon cleavage, which is useful for detecting and quantifying protease activity in biological samples like blood or tissue extracts. It is for research use only.
  • Biochemical assay reagent
  • Used as a substrate for enzymes that cleave peptide bonds (e.g., proteases)
  • Cleavage forms a yellow product (p-nitroaniline)
  • Can be used as a biological material or organic compound for life science-related research
  • Appearance: solid
  • Color: off-white to light yellow
  • Useful tool for detecting and quantifying protease activity in biological samples