Learn More
Medchemexpress LLC L-Leucyl-4-nitroanilide hydrochloride | 16010-98-3 | 100.0% | 287.74 | 250 MG
SDP
Catalog No. 5000349368
Encompass_Preferred
L-Leucyl-4-nitroanilide hydrochloride is a commonly used substrate in various biochemical assays. It helps measure the activity of enzymes that cleave peptide bonds, such as proteases.
  • Cleaved by enzymes to form a yellow product called p-nitroaniline.
  • Useful for detecting and quantifying protease activity in biological samples.
  • Can be used in blood or tissue extracts.
  • Biochemical reagent for life science related research.
Catalog No. 50-003-49368 Supplier Medchemexpress LLC Supplier No. 250MGHY137872
L-Leucyl-4-nitroanilide hydrochloride is a commonly used substrate in various biochemical assays. It helps measure the activity of enzymes that cleave peptide bonds, such as proteases.
  • Cleaved by enzymes to form a yellow product called p-nitroaniline.
  • Useful for detecting and quantifying protease activity in biological samples.
  • Can be used in blood or tissue extracts.
  • Biochemical reagent for life science related research.