Learn More
Medchemexpress LLC L-Leucyl-4-nitroanilide hydrochloride | 16010-98-3 | 99.9% | 287.74 | 10 G
SDP
Catalog No. 5000453712
Encompass_Preferred
L-Leucyl-4-nitroanilide hydrochloride is a commonly used substrate in various biochemical assays to measure the activity of enzymes that cleave peptide bonds, such as proteases. It has unique chemical properties that allow it to be cleaved by these enzymes to form a yellow product called p-nitroaniline, making it a useful tool for detecting and quantifying protease activity in biological samples like blood or tissue extracts.
  • Can be used as a biological material or organic compound for life science related research.
Catalog No. 50-004-53712 Supplier Medchemexpress LLC Supplier No. HYW01191110G
L-Leucyl-4-nitroanilide hydrochloride is a commonly used substrate in various biochemical assays to measure the activity of enzymes that cleave peptide bonds, such as proteases. It has unique chemical properties that allow it to be cleaved by these enzymes to form a yellow product called p-nitroaniline, making it a useful tool for detecting and quantifying protease activity in biological samples like blood or tissue extracts.
  • Can be used as a biological material or organic compound for life science related research.