Learn More
Medchemexpress LLC HY-129905A 5mg , L-Leucyl-L-Leucine methyl ester (hydrobromide) CAS:16689-14-8 Purity:>98%
SDP
Catalog No. 502259483
Encompass_Preferred
HY-129905A 5mg L-Leucyl-L-Leucine methyl ester (hydrobromide) CAS: 16689-14-8 L-Leucyl-L-Leucine methyl ester (LLOMe) hydrobromide, a dipeptide condensation product of L-leucine methyl ester generated by human monocytes or polymorphonuclear leukocytes, selectively eliminates lymphocytes with cytotoxic potential. L-Leucyl-L-Leucine methyl ester hydrobromide also can induce endolysosomal pathway stress. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9483 Supplier Medchemexpress LLC Supplier No. HY129905A5MG
HY-129905A 5mg L-Leucyl-L-Leucine methyl ester (hydrobromide) CAS: 16689-14-8 L-Leucyl-L-Leucine methyl ester (LLOMe) hydrobromide, a dipeptide condensation product of L-leucine methyl ester generated by human monocytes or polymorphonuclear leukocytes, selectively eliminates lymphocytes with cytotoxic potential. L-Leucyl-L-Leucine methyl ester hydrobromide also can induce endolysosomal pathway stress. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.