Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-leucylglycine | 686-50-0 | MFCD00065940 | 98.0% | 188.22 g·mol⁻¹ | C8H16N2O3 | 25 G
SDP

Catalog No. 5000303481
Encompass_Preferred

H-Leu-Gly-OH (L-leucylglycine) is a dipeptide research reagent used in biochemical and peptide studies. It is supplied as a white to off-white solid and is employed as a standard, reagent, or building block in assays and peptide synthesis. Storage recommendations and purity specifications support reliable use in research workflows.

  • Molecular weight 188.22 g·mol⁻¹.
  • Chemical formula C8H16N2O3.
  • CAS number 686-50-0.
  • Purity 98.0%.
  • Appearance white to off-white solid.
  • Intended for use in biochemical assays and peptide synthesis.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-003-03481 Supplier Medchemexpress LLC Supplier No. 25GHYW016075
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Leu-Gly-OH (L-leucylglycine) is a dipeptide research reagent used in biochemical and peptide studies. It is supplied as a white to off-white solid and is employed as a standard, reagent, or building block in assays and peptide synthesis. Storage recommendations and purity specifications support reliable use in research workflows.

  • Molecular weight 188.22 g·mol⁻¹.
  • Chemical formula C8H16N2O3.
  • CAS number 686-50-0.
  • Purity 98.0%.
  • Appearance white to off-white solid.
  • Intended for use in biochemical assays and peptide synthesis.
  • Powder storage: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.