Learn More
Medchemexpress LLC L-Linalool 1g | 126-91-0 | 1 G
SDP
Catalog No. 5000229315
Encompass_Preferred
L-linalool is the L-enantiomer of linalool, a naturally occurring monoterpene alcohol with sedative and anxiolytic properties used in neuroscience and Parkinson's disease research. It is supplied as a research-grade chemical for laboratory use.
  • Monoterpene alcohol with known sedative and anxiolytic activity.
  • Used in preclinical neuroscience and Parkinson's disease studies.
  • Molecular weight 154.25 g/mol; formula C10H18O.
  • High purity suitable for research applications.
  • Stable when stored under recommended conditions (refrigerated, inert atmosphere).
Catalog No. 50-002-29315 Supplier Medchemexpress LLC Supplier No. HY1161951G
L-linalool is the L-enantiomer of linalool, a naturally occurring monoterpene alcohol with sedative and anxiolytic properties used in neuroscience and Parkinson's disease research. It is supplied as a research-grade chemical for laboratory use.
  • Monoterpene alcohol with known sedative and anxiolytic activity.
  • Used in preclinical neuroscience and Parkinson's disease studies.
  • Molecular weight 154.25 g/mol; formula C10H18O.
  • High purity suitable for research applications.
  • Stable when stored under recommended conditions (refrigerated, inert atmosphere).