Learn More
Medchemexpress LLC L-Linalool 500mg | 126-91-0 | 154.25 g/mol | C10H18O | 500 MG
SDP
Catalog No. 5000227943
Encompass_Preferred
L-Linalool ((-)-linalool) is a naturally occurring monoterpene alcohol (C10H18O) with a molecular weight of 154.25 g/mol. It is supplied as a high-purity liquid analytical standard for laboratory research and analytical applications. L-Linalool has reported sedative and anxiolytic properties and has been used in neuropharmacology and Parkinson's disease studies. Not for human use.
  • High purity analytical standard (~98%)
  • Colorless to light yellow liquid, density 0.8657 g/cm³
  • Used in neuropharmacology and Parkinson's disease research
  • Suitable for analytical and research applications
  • Available in small laboratory pack sizes (e.g., 500 mg)
Catalog No. 50-002-27943 Supplier Medchemexpress LLC Supplier No. HY116195500MG
L-Linalool ((-)-linalool) is a naturally occurring monoterpene alcohol (C10H18O) with a molecular weight of 154.25 g/mol. It is supplied as a high-purity liquid analytical standard for laboratory research and analytical applications. L-Linalool has reported sedative and anxiolytic properties and has been used in neuropharmacology and Parkinson's disease studies. Not for human use.
  • High purity analytical standard (~98%)
  • Colorless to light yellow liquid, density 0.8657 g/cm³
  • Used in neuropharmacology and Parkinson's disease research
  • Suitable for analytical and research applications
  • Available in small laboratory pack sizes (e.g., 500 mg)