Learn More
Medchemexpress LLC L-Linalool Standard 100mg | 126-91-0 | 154.25 g/mol | C10H18O | 100 MG
SDP
Catalog No. 5000226427
Encompass_Preferred
L-Linalool (L-enantiomer of linalool, CAS 126-91-0) supplied as an analytical reference standard for research and analytical applications. The product is a colorless to light yellow liquid provided in a 100 mg package with accompanying certificate of analysis (COA) and safety data sheet (SDS).
  • Analytical reference standard for identity confirmation and quantitative assays.
  • High purity of 98.1% as stated on the COA.
  • Colorless to light yellow liquid, soluble in common organic solvents.
  • Supplied in small package sizes suitable for laboratory use, including 100 MG.
  • COA and SDS provided for lot-specific quality and safety information.
Catalog No. 50-002-26427 Supplier Medchemexpress LLC Supplier No. HY116195R100MG
L-Linalool (L-enantiomer of linalool, CAS 126-91-0) supplied as an analytical reference standard for research and analytical applications. The product is a colorless to light yellow liquid provided in a 100 mg package with accompanying certificate of analysis (COA) and safety data sheet (SDS).
  • Analytical reference standard for identity confirmation and quantitative assays.
  • High purity of 98.1% as stated on the COA.
  • Colorless to light yellow liquid, soluble in common organic solvents.
  • Supplied in small package sizes suitable for laboratory use, including 100 MG.
  • COA and SDS provided for lot-specific quality and safety information.