Learn More
Medchemexpress LLC L-Linalool Standard 250mg | 126-91-0 | 154.25 g/mol | C10H18O | 250 MG
SDP
Catalog No. 5000229795
Encompass_Preferred
L-Linalool (Standard) is an analytical reference standard of the natural monoterpene L-linalool provided for research and analytical use, including method development, chromatographic calibration, and compound identification. It is commonly used in studies of sedative and anxiolytic effects and in Parkinson's disease research.
  • Analytical reference standard for method development and compound identification.
  • High purity (98.1%) suitable for analytical applications.
  • Chemical formula C10H18O and molecular weight 154.25 g/mol.
  • Available in small laboratory pack sizes, including 250 MG.
  • Intended for research use only.
Catalog No. 50-002-29795 Supplier Medchemexpress LLC Supplier No. HY116195R250MG
L-Linalool (Standard) is an analytical reference standard of the natural monoterpene L-linalool provided for research and analytical use, including method development, chromatographic calibration, and compound identification. It is commonly used in studies of sedative and anxiolytic effects and in Parkinson's disease research.
  • Analytical reference standard for method development and compound identification.
  • High purity (98.1%) suitable for analytical applications.
  • Chemical formula C10H18O and molecular weight 154.25 g/mol.
  • Available in small laboratory pack sizes, including 250 MG.
  • Intended for research use only.