Learn More
Cayman Chemical L-Lysine lactam (hydrochloride), Purity: ≥95%, CAS: 26081-07-2, 1 g
SDP
Catalog No. 502988070
Encompass_Preferred
A building block; has been used in the synthesis of HIV protease inhibitors, as well as bengamide derivatives with in vitro anticancer activity
Catalog No. 50-298-8070 Supplier Cayman Chemical Supplier No. 316851 G
A building block; has been used in the synthesis of HIV protease inhibitors, as well as bengamide derivatives with in vitro anticancer activity