Learn More
Medchemexpress LLC L-lysinamide, l-phenylalanyl-n-[4-(hydroxymethyl)phenyl]-n6-(triphenylmethyl)- | 1116085-99-4 | 96.2% | 640.81 | C41H44N4O3 | 25 MG
SDP
Catalog No. 5000203939
Encompass_Preferred
Phe-Lys(Trt)-PAB is a cathepsin-cleavable antibody-drug conjugate (ADC) linker used in the synthesis of peptide-based ADCs. Supplied for research use, the material is provided as small laboratory pack sizes and is accompanied by a certificate of analysis documenting purity and analytical data.
  • Cathepsin-cleavable Phe-Lys linker.
  • Designed for incorporation into peptide-based ADCs.
  • Available in small milligram pack sizes, including 25 mg.
  • Purity approximately 96.2% by LCMS.
  • Provided with a certificate of analysis and handling information.
  • Intended for research use only.
Catalog No. 50-002-03939 Supplier Medchemexpress LLC Supplier No. HY12934925MG
Phe-Lys(Trt)-PAB is a cathepsin-cleavable antibody-drug conjugate (ADC) linker used in the synthesis of peptide-based ADCs. Supplied for research use, the material is provided as small laboratory pack sizes and is accompanied by a certificate of analysis documenting purity and analytical data.
  • Cathepsin-cleavable Phe-Lys linker.
  • Designed for incorporation into peptide-based ADCs.
  • Available in small milligram pack sizes, including 25 mg.
  • Purity approximately 96.2% by LCMS.
  • Provided with a certificate of analysis and handling information.
  • Intended for research use only.