Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-lysinamide, L-phenylalanyl-N-[4-(hydroxymethyl)phenyl]-N6-(triphenylmethyl) | 1116085-99-4 | 96.2% | 640.81 | C41H44N4O3 | 50 MG
SDP

Catalog No. 5000203940
Encompass_Preferred

Phe-Lys(Trt)-PAB is a cathepsin-cleavable linker used in the synthesis of antibody-drug conjugates and supplied for research use only.

  • Cathepsin-cleavable linker for ADC synthesis.
  • Suitable for designing antibody-drug conjugates with protease-triggered release.
  • Purity typically reported at 96.2% by analytical methods.
  • Molecular weight 640.81 g/mol and formula C41H44N4O3.
  • Available in small-scale research pack sizes for formulation and synthesis workflows.

Catalog No. 50-002-03940 Supplier Medchemexpress LLC Supplier No. HY12934950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phe-Lys(Trt)-PAB is a cathepsin-cleavable linker used in the synthesis of antibody-drug conjugates and supplied for research use only.

  • Cathepsin-cleavable linker for ADC synthesis.
  • Suitable for designing antibody-drug conjugates with protease-triggered release.
  • Purity typically reported at 96.2% by analytical methods.
  • Molecular weight 640.81 g/mol and formula C41H44N4O3.
  • Available in small-scale research pack sizes for formulation and synthesis workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.