Learn More
Medchemexpress LLC L-lysinamide, N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-phenylalanyl derivative | 1116086-09-9 | MFCD28898938 | >98.0% | 1028.15 g·mol^-1 | C63H57N5O9 | 50 MG
SDP
Catalog No. 5000203132
Encompass_Preferred
Fmoc-Phe-Lys(Trt)-PAB-PNP is a peptide-based, protease-cleavable linker used in the synthesis of antibody-drug conjugates (ADCs). The molecule contains an Fmoc-protected N-terminus, a para-aminobenzyl (PAB) self-immolative spacer, and a p-nitrophenyl (PNP) leaving group; it is supplied as a solid for research use only.
  • Cleavable linker designed for ADC synthesis
  • Fmoc-protected N-terminus for solid-phase peptide chemistry
  • PAB self-immolative spacer enables payload release after cleavage
  • PNP leaving group suitable for conjugation chemistry
  • Typical reported purity ≥96-98% and supplied as a solid
  • Intended for research use only; not for human or clinical use
Catalog No. 50-002-03132 Supplier Medchemexpress LLC Supplier No. HY12935050MG
Fmoc-Phe-Lys(Trt)-PAB-PNP is a peptide-based, protease-cleavable linker used in the synthesis of antibody-drug conjugates (ADCs). The molecule contains an Fmoc-protected N-terminus, a para-aminobenzyl (PAB) self-immolative spacer, and a p-nitrophenyl (PNP) leaving group; it is supplied as a solid for research use only.
  • Cleavable linker designed for ADC synthesis
  • Fmoc-protected N-terminus for solid-phase peptide chemistry
  • PAB self-immolative spacer enables payload release after cleavage
  • PNP leaving group suitable for conjugation chemistry
  • Typical reported purity ≥96-98% and supplied as a solid
  • Intended for research use only; not for human or clinical use