Learn More
Medchemexpress LLC L-lysine-1,2,3,4,5,6-13C6-N2,N6-15N2, hydrochloride | 202406-54-0 | 99.4% | 227.05 | 25 MG
SDP
Catalog No. 5000454018
Encompass_Preferred
L-Lysine-13C6,15N2 (dihydrochloride) is a 15N-labeled and 13C-labeled L-Lysine dihydrochloride. This compound is used as a tracer and an internal standard for quantitative analysis. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development, with deuteration specifically affecting pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54018 Supplier Medchemexpress LLC Supplier No. 25MGHYW009762S
L-Lysine-13C6,15N2 (dihydrochloride) is a 15N-labeled and 13C-labeled L-Lysine dihydrochloride. This compound is used as a tracer and an internal standard for quantitative analysis. Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development, with deuteration specifically affecting pharmacokinetic and metabolic profiles.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.