Learn More
Medchemexpress LLC L-lysine-13c dihydrochloride | 202190-50-9 | 220.10 | 5 MG
SDP
Catalog No. 5000390135
Encompass_Preferred
L-Lysine-13C (dihydrochloride) is the 13C-labeled L-Lysine dihydrochloride. L-lysine dihydrochloride is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health. For research use only. We do not sell to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-90135 Supplier Medchemexpress LLC Supplier No. HYN0470S25MG
L-Lysine-13C (dihydrochloride) is the 13C-labeled L-Lysine dihydrochloride. L-lysine dihydrochloride is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health. For research use only. We do not sell to patients.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.