Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-lysine-13C dihydrochloride | 202190-50-9 | 99.9% | 220.10 | C5(13C)H16Cl2N2O2 | 10 MG
SDP

Catalog No. 5000295818
Encompass_Preferred

L-lysine-13C dihydrochloride is a stable-isotope labelled form of the essential amino acid L-lysine, provided as the dihydrochloride salt for tracer, metabolic, and mass-spectrometry studies. It is 13C-labelled at the carboxyl carbon and supplied as a white to off-white solid with high chemical purity.

  • 13C-labelled at the carboxyl carbon for tracer and mass-spectrometry applications.
  • Provided as the dihydrochloride salt for improved solubility and stability.
  • High purity (99.9%) suitable for research use.
  • White to off-white solid; molecular weight 220.10 g/mol.
  • CAS number 202190-50-9 for unambiguous identification.
  • Storage: store sealed at 4°C; in solvent, store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-95818 Supplier Medchemexpress LLC Supplier No. HYN0470S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-lysine-13C dihydrochloride is a stable-isotope labelled form of the essential amino acid L-lysine, provided as the dihydrochloride salt for tracer, metabolic, and mass-spectrometry studies. It is 13C-labelled at the carboxyl carbon and supplied as a white to off-white solid with high chemical purity.

  • 13C-labelled at the carboxyl carbon for tracer and mass-spectrometry applications.
  • Provided as the dihydrochloride salt for improved solubility and stability.
  • High purity (99.9%) suitable for research use.
  • White to off-white solid; molecular weight 220.10 g/mol.
  • CAS number 202190-50-9 for unambiguous identification.
  • Storage: store sealed at 4°C; in solvent, store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.