Learn More
Medchemexpress LLC L-Lysine-13C6,15N2 dihydrochloride | 202406-54-0 | 99.6% | 227.05 | 10 MG
SDP
Catalog No. 5000452073
Encompass_Preferred
L-Lysine-13C6,15N2 (dihydrochloride) is a 15N-labeled and 13C-labled L-Lysine dihydrochloride. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • 15N-labeled and 13C-labled L-Lysine dihydrochloride
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-52073 Supplier Medchemexpress LLC Supplier No. HYW009762S10MG
L-Lysine-13C6,15N2 (dihydrochloride) is a 15N-labeled and 13C-labled L-Lysine dihydrochloride. It is for research use only and not sold to patients. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • 15N-labeled and 13C-labled L-Lysine dihydrochloride
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS