Learn More
Medchemexpress LLC L-Lysine-13C6,15N2 hydrochloride | 1200447-00-2 | 190.59 | 1 MG
SDP
Catalog No. 5000389695
Encompass_Preferred
L-Lysine-13C6,15N2 (hydrochloride) is the 13C- and 15N-labeled form of L-Lysine hydrochloride. L-lysine hydrochloride is an essential amino acid for humans with various benefits, including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses, and improving gut health. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation.
  • Deuteration has gained attention due to its potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-89695 Supplier Medchemexpress LLC Supplier No. HYN0470S31MG
L-Lysine-13C6,15N2 (hydrochloride) is the 13C- and 15N-labeled form of L-Lysine hydrochloride. L-lysine hydrochloride is an essential amino acid for humans with various benefits, including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses, and improving gut health. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation.
  • Deuteration has gained attention due to its potential to affect pharmacokinetic and metabolic profiles of drugs.