Learn More
Medchemexpress LLC L-lysine-13C6,15N2 hydrochloride | 1200447-00-2 | 190.59 | 5 MG
SDP
Catalog No. 5000389697
Encompass_Preferred
L-Lysine-13C6,15N2 (hydrochloride) is the 13C- and 15N-labeled L-Lysine hydrochloride. L-lysine hydrochloride is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Treats herpes.
  • Increases calcium absorption.
  • Reduces diabetes-related illnesses.
  • Improves gut health.
Catalog No. 50-003-89697 Supplier Medchemexpress LLC Supplier No. HYN0470S35MG
L-Lysine-13C6,15N2 (hydrochloride) is the 13C- and 15N-labeled L-Lysine hydrochloride. L-lysine hydrochloride is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Treats herpes.
  • Increases calcium absorption.
  • Reduces diabetes-related illnesses.
  • Improves gut health.