Learn More
Medchemexpress LLC L-lysine-13C6,15N2 hydrochloride | 1200447-00-2 | MFCD00144647 | 99.9% | 10 MG
SDP
Catalog No. 5000295759
Encompass_Preferred
L-lysine-13C6,15N2 hydrochloride is a stable isotope-labeled amino acid supplied as the hydrochloride salt for use in isotopic labeling experiments and quantitative proteomics. Enriched with six 13C atoms and two 15N atoms, it is provided as a white solid for laboratory research applications.
  • Provides 13C6 and 15N2 isotopic labeling for SILAC and metabolic tracing.
  • High purity suitable for mass spectrometry-based quantitation.
  • Supplied as the hydrochloride salt and dry solid for easy handling.
  • Molecular weight approximately 190.59 g/mol.
  • Store as recommended for labeled amino acids (typically at -20 °C).
Catalog No. 50-002-95759 Supplier Medchemexpress LLC Supplier No. HYN0470S310MG
Add to Cart
Add to Cart
L-lysine-13C6,15N2 hydrochloride is a stable isotope-labeled amino acid supplied as the hydrochloride salt for use in isotopic labeling experiments and quantitative proteomics. Enriched with six 13C atoms and two 15N atoms, it is provided as a white solid for laboratory research applications.
  • Provides 13C6 and 15N2 isotopic labeling for SILAC and metabolic tracing.
  • High purity suitable for mass spectrometry-based quantitation.
  • Supplied as the hydrochloride salt and dry solid for easy handling.
  • Molecular weight approximately 190.59 g/mol.
  • Store as recommended for labeled amino acids (typically at -20 °C).