Learn More
Medchemexpress LLC L-Lysine-13C6 hydrochloride | 1228077-86-8 | 188.60 | 50 MG
SDP
Catalog No. 5000388945
Encompass_Preferred
L-Lysine-13C6 hydrochloride is the 13C labeled L-Lysine hydrochloride. L-lysine hydrochloride is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily as tracers for quantitation during drug development.
Catalog No. 50-003-88945 Supplier Medchemexpress LLC Supplier No. HYN0470S850MG
L-Lysine-13C6 hydrochloride is the 13C labeled L-Lysine hydrochloride. L-lysine hydrochloride is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily as tracers for quantitation during drug development.