Learn More
Medchemexpress LLC L-Lysine-15N dihydrochloride | 204451-50-3 | > 98.0% | 220.10 | 1 MG
SDP
Catalog No. 5000451613
Encompass_Preferred
L-Lysine-15N dihydrochloride is a 15N-labeled L-Lysine dihydrochloride, primarily utilized as a tracer for quantitation in drug development processes. This compound incorporates stable heavy isotopes of hydrogen, carbon, and other elements for research applications.
  • 15N-labeled L-Lysine dihydrochloride
  • Suitable for research use only
  • Store at 4°C, sealed, and away from moisture
  • For solutions, store at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-51613 Supplier Medchemexpress LLC Supplier No. HYW009762S71MG
L-Lysine-15N dihydrochloride is a 15N-labeled L-Lysine dihydrochloride, primarily utilized as a tracer for quantitation in drug development processes. This compound incorporates stable heavy isotopes of hydrogen, carbon, and other elements for research applications.
  • 15N-labeled L-Lysine dihydrochloride
  • Suitable for research use only
  • Store at 4°C, sealed, and away from moisture
  • For solutions, store at -80°C for 6 months or -20°C for 1 month