Learn More
Medchemexpress LLC L-Lysine-15N2 (hydrochloride) | 1217460-44-0 | 184.64 | 25 MG
SDP
Catalog No. 5000389283
Encompass_Preferred
L-Lysine-15N2 hydrochloride is the 15N-labeled L-Lysine hydrochloride. It is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89283 Supplier Medchemexpress LLC Supplier No. 25MGHYN0470S
L-Lysine-15N2 hydrochloride is the 15N-labeled L-Lysine hydrochloride. It is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.