Learn More
AdipoGen L-Lysine
SDP
Supplier:  AdipoGen CDXL0110G001
Encompass_Preferred
Chemical. CAS 56-87-1. Formula C6H14N2O2. MW BD9837. L-Lysine is an essential alpha-amino acid, which is encoded by the codons AAA and AAG. It is a key building block in protein synthesis in which it acts as a base. Lysine residues are useful in many cellular processes, due to their ability to accept a wide variety of post-translational modifications. These modifications include acetylation, methylation, ubiquitination, sumoylation, neddylation, biotinylation, pupylation and carboxylation. L-lysine is an essential amino acid that cannot be synthesized by humans and mammals through transamination. Lysine plays a pivotal role in different processes inside the human body, such as proteinogenesis, catalysis, fatty acid metabolism, histone modification, polypeptide crosslinking, calcium homeostasis, and acts as a nucleophile in enzymatic reactions.
Catalog No. 50-256-0600