Learn More
Medchemexpress LLC HY-N0469 500mg Medchemexpress, L-Lysine CAS:56-87-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0469500MG
Encompass_Preferred
Medchemexpress, HY-N0469 500mg L-Lysine CAS:56-87-1 L-lysine is an essential amino acid with important roles in connective tissues and carnitine synthesis, energy production, growth in children, and maintenance of immune functions. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2792