Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lysine, 99.6%, MP Biomedicals™

Catalog No. ICN19022405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19022405 5 g
ICN19022425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19022405 Supplier MP Biomedicals, Inc Supplier No. 0219022405
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39665-12-8
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151035
InChI Key HZRUTVAFDWTKGD-XZNNNFJINA-N
Synonym l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
PubChem CID 16211825
IUPAC Name (2S)-2,6-diaminohexanoic acid hydrate
SMILES O.NCCCC[C@H](N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 164.2
Physical Form Crystalline Powder
Chemical Name or Material L-lysine, Free Base, Monohydrate
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.