Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Lysine, compd. with (aR)-a-methyl-4-(2-methylpropyl)-N-(methylsulfonyl)benzeneacetamide (1:1) | 266359-93-7 | C20H35N3O5S | 500 MG
SDP

Catalog No. 5000372412
Encompass_Preferred

Reparixin L-lysine salt is an allosteric inhibitor of chemokine receptor 1/2 (CXCR1/2) activation. It is for research use only and not sold to patients.

  • Purity: 99.87%
  • Molecular weight: 429.57
  • Molecular formula: C20H35N3O5S
  • Appearance: Solid, off-white to light yellow
  • Solubility in H2O: 100 mg/mL (232.79 mM; requires ultrasonic)
  • Solubility in DMSO: 100 mg/mL (232.79 mM; requires ultrasonic; use newly opened DMSO due to hygroscopic nature)
  • Storage: 4°C, sealed, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)

Catalog No. 50-003-72412 Supplier Medchemexpress LLC Supplier No. HY15252500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reparixin L-lysine salt is an allosteric inhibitor of chemokine receptor 1/2 (CXCR1/2) activation. It is for research use only and not sold to patients.

  • Purity: 99.87%
  • Molecular weight: 429.57
  • Molecular formula: C20H35N3O5S
  • Appearance: Solid, off-white to light yellow
  • Solubility in H2O: 100 mg/mL (232.79 mM; requires ultrasonic)
  • Solubility in DMSO: 100 mg/mL (232.79 mM; requires ultrasonic; use newly opened DMSO due to hygroscopic nature)
  • Storage: 4°C, sealed, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed, away from moisture)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.