Learn More
Medchemexpress LLC L-Lysine-d4 dihydrochloride | 203633-22-1 | 98.0% | 223.13 | 10 MG
SDP
Catalog No. 5000452134
Encompass_Preferred
L-Lysine-d4 dihydrochloride is a deuterium labeled L-Lysine dihydrochloride. It is for research use only and has not been fully validated for medical applications. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer for quantitation during drug development.
  • Can affect the pharmacokinetic and metabolic profiles of drugs.
  • Stable heavy isotope.
Catalog No. 50-004-52134 Supplier Medchemexpress LLC Supplier No. HYW009762S210MG
L-Lysine-d4 dihydrochloride is a deuterium labeled L-Lysine dihydrochloride. It is for research use only and has not been fully validated for medical applications. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Used as a tracer for quantitation during drug development.
  • Can affect the pharmacokinetic and metabolic profiles of drugs.
  • Stable heavy isotope.