Learn More
Medchemexpress LLC L-Lysine-d8 (hydrochloride) | 344298-93-7 | 190.70 | 1 MG
SDP
Catalog No. 5000389698
Encompass_Preferred
L-Lysine-d8 (hydrochloride) is the deuterium labeled L-Lysine hydrochloride. L-lysine hydrochloride is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health. It can also be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled L-Lysine hydrochloride.
  • Essential amino acid for humans.
  • Potential benefits include treating herpes, increasing calcium absorption, reducing diabetes-related illnesses, and improving gut health.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89698 Supplier Medchemexpress LLC Supplier No. HYN0470S41MG
L-Lysine-d8 (hydrochloride) is the deuterium labeled L-Lysine hydrochloride. L-lysine hydrochloride is an essential amino acid for humans with various benefits including treating herpes, increasing calcium absorption, reducing diabetes-related illnesses and improving gut health. It can also be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled L-Lysine hydrochloride.
  • Essential amino acid for humans.
  • Potential benefits include treating herpes, increasing calcium absorption, reducing diabetes-related illnesses, and improving gut health.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.