Learn More
Medchemexpress LLC L-Lysine-d8 (hydrochloride) | 344298-93-7 | 190.70 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0470S45MG
Encompass_Preferred
L-Lysine-d8 hydrochloride is a deuterium labeled form of L-Lysine hydrochloride, which is an essential amino acid for humans. This compound is valuable as a tracer in research and can serve as an internal standard for quantitative analysis in various analytical techniques.
  • Used as a tracer.
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Treats herpes.
  • Increases calcium absorption.
  • Reduces diabetes-related illnesses.
  • Improves gut health.
Catalog No. 50-003-90136