Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-lysine-d9 hydrochloride | 2708343-64-8 | 99.1% | 191.70 g/mol | C6H6D9ClN2O2 | 10 MG
SDP

Catalog No. 5000295860
Encompass_Preferred

L-Lysine-d9 hydrochloride is a deuterium-labeled form of the essential amino acid L-lysine, presented as the hydrochloride salt and intended for use as an internal standard, tracer, or reference material in mass spectrometry and metabolomics workflows.

  • Deuterium-labeled internal standard for LC-MS and metabolomics.
  • Purity approximately 99.0% (batch example 99.09%).
  • Molecular formula: C6H6D9ClN2O2.
  • Molecular weight: 191.70 g/mol.
  • Appearance: white to off-white solid.
  • Typical package: 10 MG; other sizes available.

Catalog No. 50-002-95860 Supplier Medchemexpress LLC Supplier No. HYN0470S510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Lysine-d9 hydrochloride is a deuterium-labeled form of the essential amino acid L-lysine, presented as the hydrochloride salt and intended for use as an internal standard, tracer, or reference material in mass spectrometry and metabolomics workflows.

  • Deuterium-labeled internal standard for LC-MS and metabolomics.
  • Purity approximately 99.0% (batch example 99.09%).
  • Molecular formula: C6H6D9ClN2O2.
  • Molecular weight: 191.70 g/mol.
  • Appearance: white to off-white solid.
  • Typical package: 10 MG; other sizes available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.