Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-Lysine ethyl ester diisocyanate, 97%

Click to view available options
Quantity:
5g
25g
45172-15-4
C10H14N2O4
226.232
MFCD08276480
IGWTZHMYJZZIGC-VIFPVBQESA-N
s-ethyl 2,6-diisocyanatohexanoate,l-lysine ethyl ester diisocyanate,ethyllysine diisocyanate,ethyllysine diisocyanate;,ksc005q3n,lysine ethyl ester diisocyanate;,l-lysine diisocyanate ethyl ester,l-lysine ethyl ester diisocyanate;,ethyl 2s-2,6-diisocyanatohexanoate,s-2,6-diisocyanatohexanoic acid ethyl ester
71440643
ethyl (2S)-2,6-diisocyanatohexanoate
CCOC(=O)C(CCCCN=C=O)N=C=O
45172-15-4
C10H14N2O4
226.232
MFCD08276480
IGWTZHMYJZZIGC-VIFPVBQESA-N
s-ethyl 2,6-diisocyanatohexanoate,l-lysine ethyl ester diisocyanate,ethyllysine diisocyanate,ethyllysine diisocyanate;,ksc005q3n,lysine ethyl ester diisocyanate;,l-lysine diisocyanate ethyl ester,l-lysine ethyl ester diisocyanate;,ethyl 2s-2,6-diisocyanatohexanoate,s-2,6-diisocyanatohexanoic acid ethyl ester
71440643
ethyl (2S)-2,6-diisocyanatohexanoate
CCOC(=O)C(CCCCN=C=O)N=C=O
Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Sparingly soluble in water (0.19 g/L).

Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
Refractive Index 1.4572
Formula Weight 226.23
Physical Form Viscous Liquid
Percent Purity 97%
Color Yellow to Brown
Quantity 5g
Sensitivity Moisture Sensitive
Chemical Name or Material L-Lysine ethyl ester diisocyanate

GHS H Statement
H331-H334-H315-H319-H317-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P285-P261-P280-P305+P351+P338-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : 2° to 8°C