Learn More
Research Products International Corp L-Lysine Hydrochloride, 500 Grams
SDP
Supplier:  Research Products International Corp L37040500.0
Encompass_Preferred
L-Lysine is a fundamental building block for protein synthesis. It is incorporated into polypeptide chains during translation, contributing to the structure and function of proteins within cells. 
L-Lysine Hydrochloride is utilized in laboratory research, particularly in studies related to biochemistry, molecular biology, and cell biology. 
Its role as an essential amino acid makes it valuable for understanding cellular processes.
Catalog No. 50-213-414