Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Lysine, Hydrate, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18611172 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-611-172 5 kg Amber Glass Bottle
18-611-171 5 g Amber Glass Bottle
18-611-170 25 g Amber Glass Bottle
18-611-169 100 g Amber Glass Bottle
18-611-168 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-611-172 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L11465KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

CAS: 39665-12-8 | 164.21 g/mol

Chemical Identifiers

CAS 39665-12-8

Specifications

Ammonium 200 ppm
CAS Max % 1
pH 9.0 to 10.5
Chloride 300 ppm
Heavy Metals 10 ppm
Iron (Fe) 10 ppm
Other Amino Acids 0.005
Packaging Amber Glass Bottle
Specific Rotation +25.5° to+27.0°
Sulfate 200 ppm
Water 10.0 - 11.5%
Molecular Weight (g/mol) 164.21
Quantity 5 kg
Percent Purity 98.5 to 101.5%
Grade Reagent
Chemical Name or Material L-(+)-Lysine Hydrate
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.