Learn More
Alkali Scientific L Lysine Hydrochloride, 50 Grams
SDP
Catalog No. 5000661856
Encompass_Preferred
L-Lysine Hydrochloride is a high-purity amino acid available in a 50-gram package. This compound is essential in protein synthesis and is commonly used in research on enzyme activity, metabolic pathways, and nutritional studies. L-Lysine plays a critical role in cell growth and repair, as well as in the production of collagen and other structural proteins. It is frequently used in laboratory cultures and as a supplement in cell media to promote cell health and development. The powder form ensures easy handling and precise measurement, and the high purity guarantees reliable results in analytical and biochemical applications.
Catalog No. 50-006-61856 Supplier Alkali Scientific Supplier No. LLY004CP
L-Lysine Hydrochloride is a high-purity amino acid available in a 50-gram package. This compound is essential in protein synthesis and is commonly used in research on enzyme activity, metabolic pathways, and nutritional studies. L-Lysine plays a critical role in cell growth and repair, as well as in the production of collagen and other structural proteins. It is frequently used in laboratory cultures and as a supplement in cell media to promote cell health and development. The powder form ensures easy handling and precise measurement, and the high purity guarantees reliable results in analytical and biochemical applications.