Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Lysine Hydrochloride, 99+%, MP Biomedicals

Catalog No. ICN19469780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19469780 100 g
ICN19469790 500 g
ICN19469791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19469780 Supplier MP Biomedicals, Inc Supplier No. 0219469780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 657-27-2 | C6H15ClN2O2 | 182.65 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00064564 MFCD00081870
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
IUPAC Name (2S)-2,6-diaminohexanoic acid hydrochloride
SMILES Cl.NCCCC[C@H](N)C(O)=O

Specifications

Quantity 100 g
Formula Weight 182.6
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Lysine, 99+%, Hydrochloride
Recommended Storage Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.