Learn More
Medchemexpress LLC L-lysine, L-lysyl-L-glutaminyl-L-phenylalanyl- | 99.6% | 891.73 | 5 MG
SDP
Catalog No. 5000400161
Encompass_Preferred
KQFK TFA is an inactive control peptide, supplied as the TFA salt form. This compound, with a molecular formula of C32H46F9N7O12 and a molecular weight of 891.73, presents as a white to off-white solid with a purity of 99.6%. It is intended for research use only.
  • Inactive control peptide, TFA salt form
  • White to off-white solid appearance
  • High purity of 99.6%
  • Powder can be stored for 2 years at -80°C or 1 year at -20°C in sealed conditions, away from moisture
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C with sealed storage, away from moisture
Catalog No. 50-004-00161 Supplier Medchemexpress LLC Supplier No. HYP3970D5MG
KQFK TFA is an inactive control peptide, supplied as the TFA salt form. This compound, with a molecular formula of C32H46F9N7O12 and a molecular weight of 891.73, presents as a white to off-white solid with a purity of 99.6%. It is intended for research use only.
  • Inactive control peptide, TFA salt form
  • White to off-white solid appearance
  • High purity of 99.6%
  • Powder can be stored for 2 years at -80°C or 1 year at -20°C in sealed conditions, away from moisture
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C with sealed storage, away from moisture