Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-lysine, L-valyl-L-glutaminyl-L-isoleucyl-L-valyl-L-tyrosyl- (TFA) | 99.45% | 748.91 | 25 MG
SDP

Catalog No. 5000400382
Encompass_Preferred

PHF6 TFA is a self-assembly sequence capable of initiating the full-length tau protein aggregation, mapped to the third microtubule-binding repeat region of the tau protein. Aggregated PHF6 TFA potentiates NLRP3 inflammasome expression and autophagy in human microglial cells.

  • Purity: 99.45%
  • Molecular weight: 748.91 (free base)
  • Chemical formula: C36H60N8O9.xC2HF3O2
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Val-Gln-Ile-Val-Tyr-Lys
  • Solubility in H2O: ≥ 50 mg/mL
  • Store sealed and away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-00382 Supplier Medchemexpress LLC Supplier No. 25MGHYP4808A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PHF6 TFA is a self-assembly sequence capable of initiating the full-length tau protein aggregation, mapped to the third microtubule-binding repeat region of the tau protein. Aggregated PHF6 TFA potentiates NLRP3 inflammasome expression and autophagy in human microglial cells.

  • Purity: 99.45%
  • Molecular weight: 748.91 (free base)
  • Chemical formula: C36H60N8O9.xC2HF3O2
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Val-Gln-Ile-Val-Tyr-Lys
  • Solubility in H2O: ≥ 50 mg/mL
  • Store sealed and away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.