Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L(+)-Lysine monohydrate, 99%

Catalog No. AC173735000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173735000 500 g Plastic Bottle
AC173730250 25 g Plastic Bottle
AC173731000 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC173735000 Supplier Thermo Scientific Chemicals Supplier No. 173735000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 39665-12-8 | C6H16N2O3 | 164.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39665-12-8
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151035
InChI Key HZRUTVAFDWTKGD-XZNNNFJINA-N
Synonym l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
PubChem CID 16211825
SMILES O.NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 212.0°C to 214.0°C
Ammonium 200ppm max.
pH 9 to 10.5 (2.5% soln. at 25°C)
Color White to Yellow
Chlorine (Cl) 300ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Bottle
Linear Formula H2N(CH2)4CH(NH2)CO2H·H2O
Specific Rotation + 26.25
Beilstein 04, 435
Sulfate 200ppm max.
Merck Index 14, 5636
Transmittance 95% min. (c=5, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 1500g/L (20°C). Other solubilities: insoluble in common neutral solvents
Water 10 to 11.5%
Specific Rotation Condition + 26.25 (20.00°C c=8,6N HCl)
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrate
Quantity 500 g
Formula Weight 164.21
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Lysine monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.