Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-lysine, N6-[(1,1-dimethylethoxy)carbonyl]-, phenylmethyl ester, hydrochloride (1:1) | 133170-57-7 | MFCD11045868 | 97.5% | 372.89 | C18H29ClN2O4 | 100 G
SDP

Catalog No. 5000208294
Encompass_Preferred

Benzyl N6-(t-Boc)-L-lysinate hydrochloride is a Boc-protected L-lysine derivative used as a building block in peptide synthesis and organic synthesis. It is supplied as a research-grade biochemical reagent for laboratory use.

  • Protected lysine derivative suitable for peptide synthesis.
  • Cas number 133170-57-7.
  • Molecular formula C18H29ClN2O4; molecular weight 372.89.
  • Purity (LCMS) 97.49%.
  • Storage: store at room temperature; keep dry and cool.
  • Available in laboratory quantities up to 100 g.

Catalog No. 50-002-08294 Supplier Medchemexpress LLC Supplier No. HYW088712100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Benzyl N6-(t-Boc)-L-lysinate hydrochloride is a Boc-protected L-lysine derivative used as a building block in peptide synthesis and organic synthesis. It is supplied as a research-grade biochemical reagent for laboratory use.

  • Protected lysine derivative suitable for peptide synthesis.
  • Cas number 133170-57-7.
  • Molecular formula C18H29ClN2O4; molecular weight 372.89.
  • Purity (LCMS) 97.49%.
  • Storage: store at room temperature; keep dry and cool.
  • Available in laboratory quantities up to 100 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.