Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-lysine, N6-(1-oxo-2-buten-1-yl)- | 1338823-35-0 | MFCD22381072 | 99.4% | 214.26 | C10H18N2O3 | 100 MG
SDP

Catalog No. 5000210895
Encompass_Preferred

N6-(But-2-enoyl)-L-lysine is a lysine derivative supplied as a white to off-white solid for research use in biochemical and pharmacological studies. It has been characterized by CAS number, molecular formula, and molecular weight, and is provided with a certificate of analysis to support its stated purity and storage recommendations.

  • chemical identity: N6-(but-2-enoyl)-L-lysine, CAS 1338823-35-0.
  • molecular formula: C10H18N2O3; molecular weight: 214.26 g/mol.
  • appearance: white to off-white solid; powder form for easy handling.
  • purity: high purity suitable for research applications (reported 99.43%).
  • storage: powder: -20°C for long-term (3 years) or 4°C for short-term (2 years); in solvent: -80°C for 6 months.
  • solubility: soluble in water (approx. 17.5 mg/mL with ultrasonication).
  • packaging: supplied in research-scale quantities (e.g., 100 mg) with COA documentation.

Catalog No. 50-002-10895 Supplier Medchemexpress LLC Supplier No. HYW048203100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N6-(But-2-enoyl)-L-lysine is a lysine derivative supplied as a white to off-white solid for research use in biochemical and pharmacological studies. It has been characterized by CAS number, molecular formula, and molecular weight, and is provided with a certificate of analysis to support its stated purity and storage recommendations.

  • chemical identity: N6-(but-2-enoyl)-L-lysine, CAS 1338823-35-0.
  • molecular formula: C10H18N2O3; molecular weight: 214.26 g/mol.
  • appearance: white to off-white solid; powder form for easy handling.
  • purity: high purity suitable for research applications (reported 99.43%).
  • storage: powder: -20°C for long-term (3 years) or 4°C for short-term (2 years); in solvent: -80°C for 6 months.
  • solubility: soluble in water (approx. 17.5 mg/mL with ultrasonication).
  • packaging: supplied in research-scale quantities (e.g., 100 mg) with COA documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.